Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Artemether
GREENER_CHOICESDP

Catalog No. 681850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-185-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 68-185-0 Supplier Tocris Bioscience™ Supplier No. 6818/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Prevents cerebral malaria in Plasmodium berghei infected mice.

Artemether is an antimalarial. Prevents cerebral malaria in Plasmodium berghei infected mice. Exhibits synergistic effects with Methylene Blue (Cat. No. 3213) against P. falciparum in vitro. Inhibits neuroinflammation in neuronal/microglial cell co-cultures via induction of Nrf2 expression.

Specifications

Chemical Name or Material (3R,5aS,6R,8aS,9R,10S,12R,12aR)-Decahydro-10-methoxy-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin
CAS 71963-77-4
Quantity 50 mg
Molecular Formula C16H26O5
Formula Weight 298.37
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.