Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Artemin Antibody (RM0074-6G32), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121701AF700
Description
Artemin Monoclonal specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Artemin | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ARTN | |
Mouse recombinant Artemin | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Artemin. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
RM0074-6G32 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin | |
Rat | |
Protein A or G purified | |
Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience | |
9048 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction