Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Artemin Antibody (RM0074-6G32), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121701PE
Description
Artemin Monoclonal antibody specifically detects Artemin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
| Artemin | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| RM0074-6G32 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
| artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin | |
| Mouse recombinant Artemin | |
| 0.1 mL | |
| Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience | |
| 9048 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction