Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Artemin Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | Artemin |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5118963
|
Invitrogen™
PA5118963 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5122227
|
Invitrogen™
PA5122227 |
20 μL |
Each for $237.00
|
|
|||||
Description
Artemin Polyclonal Antibody for Western Blot
The protein encoded by this gene is a member of the glial cell line-derived neurotophic factor (GDNF) family of ligands which are a group of ligands within the TGF-beta superfamily of signaling molecules. GDNFs are unique in having neurotrophic properties and have potential use for gene therapy in neurodegenerative disease. Artemin has been shown in culture to support the survival of a number of peripheral neuron populations and at least one population of dopaminergic CNS neurons. Its role in the PNS and CNS is further substantiated by its expression pattern in the proximity of these neurons. This protein is a ligand for the RET receptor and uses GFR-alpha 3 as a coreceptor. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
Artemin | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Rat | |
ARTN | |
Artemin; ARTN; Enovin; EVN; NBN; neublastin; neurotrophic factor | |
ARTN | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q6AYE8 | |
362572 | |
Recombinant protein Artemin. The antigen corresponds to amino acid range 112-224 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title