Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Artificial Blood
SDPSCI_ED

Catalog No. p-7225677 Shop All 3B Scientific Products
Click to view available options
Size:
1 L
1 gal.
This item is not returnable. View return policy
This item is not returnable. View return policy

Although not the same viscosity as real blood, Life/form™ simulation blood provides a sense of reality to any activity.

  • Although not the same viscosity as real blood, Life/form™ simulation blood provides a sense of reality to any activity.
  • For use with Life/form™ Products:
    • Advanced Venipuncture and Injection Arm (W44216, W44217)
    • Venipuncture and Injection Demonstration Arm
    • Deluxe IV Training Arm
    • Venipuncture Training Arm
    • Pediatric Arm (W44715)
    • Pediatric Head (W44101)
    • Central Venous Cannulation Simulator (W44017)
    • Heart Catherization Simulator for Total Parenteral Nutrition and Central Venous Catheterization (W44015)
    • First Aid Arm (W44733)

Specifications

Product Type Artificial Blood
Color Red
For Use With (Equipment) Life/Form™ Advanced Venipuncture and Injection Arm, Venipuncture and Injection Demonstration Arm, Deluxe IV Training Arm, Venipuncture Training Arm, Pediatric Arm, Pediatric Head, Central Venous Cannulation Simulator, Heart Catherization Simulator for Total Parenteral Nutrition and Central Venous Catheterization, First Aid Arm
Size 1 L
Weight (English) 0.18 lb.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.