Learn More
Invitrogen™ ARTS Polyclonal Antibody

Description
In Western blot applications, this antibody detects a band at ~32kDa. A suggested positive control is human lung tissue lysate. PA5-20001 can be used with blocking peptide PEP-0123.
Specifications
Specifications
| Antigen | ARTS |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SEPTIN4 |
| Gene Accession No. | O43236 |
| Gene Alias | 4-Sep; apoptosis-related protein in the TGF-beta signaling pathway; ARTS; Bh5; BRADEION; Bradeion beta; brain protein H5; C17orf47; CE5B3; CE5B3 beta; cell division control-related protein 2; cell division control-related protein 2b; cerebral protein 7; EG3-1RVC; EG3RVC; expression gene 3 in rat visual cortex; expression gene 3-1 in rat visual cortex; H5; hCDCREL-2; hucep-7; MART; M-Septin; OTTMUSG00000001265; peanut-like 2; peanut-like 2 homolog; peanut-like protein 2; PNUTL2; SEP4; Sept4; septin 4; septin H5; Septin4; septin-4; septin-M; Uncharacterized protein C17orf47 |
| Gene Symbols | SEPTIN4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.