Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARV1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113911 Supplier Invitrogen™ Supplier No. PA5113911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QAIRVTLNIN RKLSFLAVLS GLLLESIMVY FFQSMEWDVG SDYAIFKSQD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 85%; Dog: 79%; Goat: 85%; Human: 100%; Pig: 85%; Rabbit: 79%.

May act as a mediator of sterol homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARV1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ARV1
Gene Accession No. Q9H2C2
Gene Alias 1110067L22Rik; AI461928; Arv1; ARV1 fatty acid homeostatsis modulator; ARV1 homolog (S. cerevisiae); ARV1 homolog (yeast); ARV1 homolog, fatty acid homeostasis modulator; ARV1 homolog, fatty acid homeostatsis modulator; AW121084; hARV1; HT035; Protein ARV1; RGD1310264
Gene Symbols ARV1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ARV1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64801
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.