Learn More
Invitrogen™ ARVCF Polyclonal Antibody

Description
Immunogen sequence: DDTRSLAADD EGGPELEPDY GTATRRRPEC GRGLHTRAYE DTADDGGELA DERPAFPMVT AP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
Specifications
Specifications
| Antigen | ARVCF |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ARVCF |
| Gene Accession No. | O00192 |
| Gene Alias | armadillo repeat gene deleted in velocardiofacial syndrome; armadillo repeat gene deleted in velo-cardio-facial syndrome; armadillo repeat protein deleted in velo-cardio-facial syndrome; Armadillo repeat protein deleted in velo-cardio-facial syndrome homolog; Arvcf |
| Gene Symbols | ARVCF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.