Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARVCF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565933
Description
Immunogen sequence: HVALQLERAQQ PGMVSGGMGS GQPLPMAWQQ LVLQEQSPGS QASLATMPEA PDVLEETVTV EEDPGTPTSH VSIV Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
Involved in protein-protein interactions at adherens junctions.
Specifications
ARVCF | |
Polyclonal | |
Unconjugated | |
ARVCF | |
armadillo repeat gene deleted in velocardiofacial syndrome; armadillo repeat gene deleted in velo-cardio-facial syndrome; armadillo repeat protein deleted in velo-cardio-facial syndrome; Armadillo repeat protein deleted in velo-cardio-facial syndrome homolog; Arvcf | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
421 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00192 | |
ARVCF | |
Recombinant Human ARVCF. Recombinant protein control fragment (Product #RP-106615). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction