Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARX Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106787
Description
Antibody detects endogenous levels of total ARX.
Arx contains two conserved domains, a C-peptide (or aristaless domain) and the prd-like class homeobox domain. It is a member of the group-II aristaless-related protein family whose members are expressed primarily in the central and/or peripheral nervous system. This protein is thought to be involved in CNS development. Mutations in the Arx gene cause X-linked mental retardation and epilepsy.
Specifications
| ARX | |
| Polyclonal | |
| Unconjugated | |
| ARX | |
| aristaless related homeobox; aristaless-related homeobox; aristaless-related homeobox, X-linked; ARX; arx {ECO:0000250; cancer/testis antigen 121; CT121; EIEE1; Homeobox protein ARX; ISSX; MRX29; MRX32; MRX33; MRX36; MRX38; MRX43; MRX54; MRX76; MRX87; MRXS1; PRTS; RGD1562672; UniProtKB:Q96QS3} | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11878, 170302, 317268 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A6YP92, O35085, Q96QS3 | |
| ARX | |
| A synthesized peptide derived from human ARX(Accession Q96QS3), corresponding to amino acid residues A278-Q328. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction