Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arylsulfatase A/ARSA Antibody (001), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290354AF750
Description
Arylsulfatase A/ARSA Monoclonal specifically detects Arylsulfatase A/ARSA in Mouse samples. It is validated for ELISA.Specifications
Arylsulfatase A/ARSA | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human ARSA | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
1 | |
ELISA | |
arylsulfatase A, ASA, Cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Arylsulfatase A/ARSA (NP_033843.2; Met 1-Ser 506). | |
0.1 mL | |
Lipid and Metabolism | |
410 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction