Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arylsulfatase A/ARSA Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298210AF700
Description
Arylsulfatase A/ARSA Polyclonal specifically detects Arylsulfatase A/ARSA in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Arylsulfatase A/ARSA | |
Polyclonal | |
Western Blot, ELISA | |
arylsulfatase A, ASA, Cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD | |
Produced in rabbits immunized with purified, recombinant Arylsulfatase A/ARSA (NP_033843.2; Met 1-Ser 506). | |
0.1 mL | |
Lipid and Metabolism | |
410 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction