Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arylsulfatase A/ARSA Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299307APC
Description
Arylsulfatase A/ARSA Polyclonal antibody specifically detects Arylsulfatase A/ARSA in Human samples. It is validated for Western Blot, ELISASpecifications
Arylsulfatase A/ARSA | |
Polyclonal | |
Western Blot, ELISA | |
arylsulfatase A, ASA, Cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD | |
Produced in rabbits immunized with purified, recombinant Human Arylsulfatase A/ARSA (Uniprot#: P15289-1; Met 1-Ala 507). | |
0.1 mL | |
Lipid and Metabolism | |
410 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction