Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Arylsulfatase A Monoclonal Antibody (498CT24.1.2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537586
Description
Arylsulfatase A Monoclonal Antibody for Western Blot
The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy, a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.
Specifications
Arylsulfatase A | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P15289 | |
ARSA | |
KLH conjugated synthetic peptide between 408-439 amino acids from the C-terminal region of human ARSA. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
498CT24.1.2 | |
Unconjugated | |
ARSA | |
ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40 | |
Mouse | |
Protein G | |
RUO | |
410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction