Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arylsulfatase A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547330
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547330 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547330 Supplier Invitrogen™ Supplier No. PA547330
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 50% cross-reactivity with recombinant mouse ARSA is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy, a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arylsulfatase A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ARSA
Gene Accession No. P15289
Gene Alias ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40
Gene Symbols ARSA
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human ARSA Arg19-Ala507.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 410
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.