Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Arylsulfatase A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586537
Description
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy, a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.
Specifications
Arylsulfatase A | |
Polyclonal | |
Unconjugated | |
ARSA | |
ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
315222, 410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P15289 | |
ARSA | |
Synthetic peptide, corresponding to amino acids 260-310 of Human ARSA. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction