Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arylsulfatase B/ARSB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215474
Description
Arylsulfatase B/ARSB Polyclonal specifically detects Arylsulfatase B/ARSB in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Arylsulfatase B/ARSB | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
arylsulfatase B, ASB, EC 3.1.6.12, G4S, MPS6, N-acetylgalactosamine-4-sulfatase | |
Rabbit | |
60 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Cat (89%), Dog (86%), Bovine (90%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
6.11mg/mL | |
Western Blot 1:500-1:3000 | |
P15848 | |
ARSB | |
Recombinant protein encompassing a sequence within the center region of human ARSB. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
411 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction