Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arylsulfatase B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111086
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111086 Supplier Invitrogen™ Supplier No. PA5111086
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TRCALDFRDG EEVATGYKNM YSTNIFTKRA IALITNHPPE KPLFLYLALQ SVHEPLQVPE EYLKPYDFIQ DKNRHHYAGM VSLMDEAVGN VT Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 80%.

Arylsulfatase B encoded by this gene belongs to the sulfatase family. The arylsulfatase B homodimer hydrolyzes sulfate groups of N-Acetyl-D-galactosamine, chondriotin sulfate, and dermatan sulfate. The protein is targetted to the lysozyme. Mucopolysaccharidosis type VI is an autosomal recessive lysosomal storage disorder resulting from a deficiency of arylsulfatase B. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arylsulfatase B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARSB
Gene Accession No. P15848
Gene Alias 1110007C02Rik; AI480648; Arsb; Arylsulfatase B; Arylsulfatase B (MPS VI); arylsulfatase B complex; arylsulfatase B regulation; arylsulfatase B structural; arylsulfatase B temporal regulation; As1; As-1; As-1r; As1-r; As-1s; As1-s; As-1t; As1-t; ASB; Asr-1; Ast-1; G4S; MPS6; N-acetylgalactosamine-4-sulfatase
Gene Symbols ARSB
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Arylsulfatase B. Recombinant protein control fragment (Product #RP-96819).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 411
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.