Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arylsulfatase D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552087 Supplier Invitrogen™ Supplier No. PA552087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGAEARSAHE FLFHYCGQHL HAARWHQKDS GSVWKVHYTT PQFHPEGAGA CYGRGVCPCS GEGVTHHRPP LLFDLSRDPS EARPLTPDSE PLYHAVIARV GAAVSEHRQT LSPVPQQFSM SNILWKPWLQ PCCGHFPFCS CHED Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 50%.

The protein encoded by this gene is a member of the sulfatase family. Sulfatases are essential for the correct composition of bone and cartilage matrix. This encoded protein is postranslationally glycosylated and localized to the lysosome. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arylsulfatase D
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARSD
Gene Accession No. P51689
Gene Alias ARSD; Arylsulfatase D; ASD; testis tissue sperm-binding protein Li 39a
Gene Symbols ARSD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Arylsulfatase D. Recombinant protein control fragment (Product #RP-92109).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.