Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arylsulfatase E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590593
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590593 Supplier Invitrogen™ Supplier No. PA590593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSETHILTPA SEPVFYQVME RVQQAVWEHQ RTLSPVPLQL DRLGNIWRPW LQPCCGPFPL CWCLREDDPQ; Positive Samples: Mouse liver, Rat liver; Cellular Location: Golgi apparatus, Golgi stack.

Arylsulfatase E is a member of the sulfatase family. It is glycosylated postranslationally and localized to the golgi apparatus. Sulfatases are essential for the correct composition of bone and cartilage matrix. X-linked chondrodysplasia punctata, a disease characterized by abnormalities in cartilage and bone development, has been linked to mutations in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arylsulfatase E
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Arse
Gene Alias ARSE; Arylsulfatase E; arylsulfatase E (chondrodysplasia punctata 1); ASE; CDPX; CDPX1; CDPXR
Gene Symbols Arse
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 520-589 of human ARSE (NP_0000382).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310326
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.