Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arylsulfatase F Polyclonal Antibody
Arylsulfatase F Polyclonal Antibody
Supplier: Thermo Scientific PA5101436
Description
Arylsulfatase F Polyclonal Antibody for Western Blot, ICC/IF

Specifications
Arylsulfatase F | |
Polyclonal | |
Unconjugated | |
ARSF | |
ARSF; Arylsulfatase F; ASF | |
Rabbit | |
Affinity Chromatography | |
RUO | |
416 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P54793 | |
ARSF | |
A synthesized peptide derived from human ARSF(Accession P54793), corresponding to amino acid residues G309-L359. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction