Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
AS 1517499 is a potent STAT6 inhibitor (IC50 = 21 nM). In vitro, it inhibits IL-4-induced Th2 differentiation of mouse spleen T cells (IC50 = 2.3 nM), without influencing Th1 differentiation induced by IL-12. In a mouse model of allergic bronchial asthma, treatment with AS 1517499 reduces mouse airway IL-3 production and partly inhibits the RhoA upregulation in bronchial smooth muscles.
Specifications
Specifications
| Chemical Name or Material | 2-[[2-(3-Chloro-4-hydroxyphenyl)ethyl]amino]-4-[(phenylmethyl)amino]-5-pyrimidinecarboxamide |
| CAS | 919486-40-1 |
| Quantity | 50 mg |
| Target | STAT6 |
| Molecular Formula | C20H20ClN5O2 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

