Learn More
AS160 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303729A

Description
The recommended shelf life for this product is 1 year from date of receipt.
AS160 (TBC1D4) is a GTPase-activiting protein for RAB2A, RAB8A, RAB10, and RAB14. The isoform 2 version of this protein promotes the insulin-induced glucose transporter SLC2A4/GLUT4 translocation at the plasma membrane. This movement increases glucose uptake. Mutations associated with the gene cause Diabetes mellitus, non-insulin-dependent, 5 (NIDDM5).Specifications
AS160 | |
Polyclonal | |
Unconjugated | |
TBC1D4 | |
Acrg embryonic lethality minimal region ortholog; TBC (Tre-2, BUB2, CDC16) domain-containing protein; TBC1 domain family member 4; akt substrate of 160 kDa | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9882 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O60343 | |
TBC1D4 | |
Between 1248 and 1298 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.