Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AS2717638
GREENER_CHOICESDP

Catalog No. 773010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-301-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 77-301-0 Supplier Tocris Bioscience™ Supplier No. 7730/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective, brain penetrant LPA5 antagonist

AS2717638 is a potent LPA5 receptor antagonist (IC50 = 38 nM in BV-2 microglia cells). It is selective for LPA5 over LPA1, LPA2, and LPA3. It inhibits LPA-mediated pro-inflammatory transcription factor phosphorylation. In vivo, AS2717638 inhibits LPA5 agonist-induced allodynia in mice. It also exhibits analgesic effects against both neuropathic and inflammatory pain in rodent models. Orally bioavailable and brain penetrant.

Specifications

Chemical Name or Material 6,7-Dimethoxy-2-(5-methyl-1,2-benzisoxazol-3-yl)-4-(1-piperidinylcarbonyl)-1(2 H)-isoquinolinone
CAS 2148339-28-8
Quantity 10 mg
Target LPAR5/LPA5/GPR92
Molecular Formula C25H25N3O5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.