Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AS3MT Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297425G
Description
AS3MT Polyclonal antibody specifically detects AS3MT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
AS3MT | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
arsenic (+3 oxidation state) methyltransferase, arsenite methyltransferase, CYT19methyltransferase cyt19, EC 2.1.1.137, Methylarsonite methyltransferase, S-adenosyl-L-methionine:arsenic(III) methyltransferase, S-adenosylmethionine:arsenic (III) methyltransferase | |
Produced in rabbits immunized with E. coli-derived Human AS3MT fragment. | |
0.1 mL | |
Lipid and Metabolism | |
57412 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction