Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASAH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109416 Supplier Invitrogen™ Supplier No. PA5109416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IVAEDKKGHL IHGRNMDFGV FLGWNINNDT WVITEQLKPL TVNLDFQRNN KTVFKASSFA GYVGMLTGFK PGLFSLTLNE RFSINGGYLG ILEWILGKKD VMWIGFLTRT VLENSTSYEE AKNLLTKTKI LAPAYFILGG NQSGEGCVIT RDRKESLDVY ELDAKQGRWY VVQTNYDRWK HPFFLDDRRT PAKMCLNRTS QENISFETMY DVLSTKPVLN KLTVYTTLID VTKGQFETYL RDCPDPCIGW.

ASAH1 encodes a member of the acid ceramidase family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed. The processing of this preproprotein generates alpha and beta subunits that heterodimerize to form the mature lysosomal enzyme, which catalyzes the degradation of ceramide into sphingosine and free fatty acid. This enzyme is overexpressed in multiple human cancers and may play a role in cancer progression. Mutations in this gene have been linked to the lysosomal storage disorder, Farber lipogranulomatosis, and a neuromuscular disorder, spinal muscular atrophy with progressive myoclonic epilepsy. The related pathways include Innate Immune System and Sphingolipid metabolism. Diseases associated with ASAH1 include Farber Lipogranulomatosis and Spinal Muscular Atrophy With Progressive Myoclonic Epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASAH1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ASAH1
Gene Accession No. Q6P7S1, Q9WV54
Gene Alias 2310081N20Rik; AC; ACDase; acid CDase; acid ceramidase; Acid ceramidase subunit alpha; Acid ceramidase subunit beta; Acylsphingosine deacylase; ASAH; Asah1; HSD33; HSD-33; N-acylethanolamine hydrolase ASAH1; N-acylsphingosine amidohydrolase; N-acylsphingosine amidohydrolase (acid ceramidase) 1; N-acylsphingosine amidohydrolase 1; PHP; PHP32; putative 32 kDa heart protein; SMAPME
Gene Symbols ASAH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 140-389 of human ASAH1 (NP_001120977.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11886, 84431
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.