Learn More
Invitrogen™ ASAHL Monoclonal Antibody (511702)

Description
In direct ELISAs, 100% cross-reactivity with recombinant mouse ASAHL is observed and no cross-reactivity with recombinant human ASAH2 or recombinant mouse ASAH2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | ASAHL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 511702 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Naaa |
| Gene Accession No. | Q02083 |
| Gene Alias | 2210023K21Rik; 3830414F09Rik; acid ceramidase-like protein; Asahl; ASAH-like protein; LOW QUALITY PROTEIN: N-acylethanolamine-hydrolyzing acid amidase; Naaa; N-acylethanolamine acid amidase; N-acylethanolamine-hydrolyzing acid amidase; N-acylethanolamine-hydrolyzing acid amidase subunit alpha; N-acylethanolamine-hydrolyzing acid amidase subunit beta; N-acylsphingosine amidohydrolase-like; PLT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.