Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Antibody, Novus Biologicals™
SDP

Catalog No. p-7108548 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP162534 100 μL
NBP16253420 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP162534 Supplier Novus Biologicals Supplier No. NBP162534
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase Polyclonal specifically detects ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ASAHL/N-acylethanolamine-hydrolyzing Acid Amidase
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q02083
Gene Alias Acid ceramidase-like protein, ASAHL, ASAH-like protein, EC 3.5.1.-, N-acylethanolamine acid amidase, N-acylethanolamine-hydrolyzing acid amidase, N-acylsphingosine amidohydrolase (acid ceramidase)-like, N-acylsphingosine amidohydrolase-like, PLT
Gene Symbols NAAA
Host Species Rabbit
Immunogen Synthetic peptides corresponding to ASAHL The peptide sequence was selected from the middle region of ASAHL. Peptide sequence VSWLIRATLSESENFEAAVGKLAKTPLIADVYYIVGGTSPREGVVITRNR.
Molecular Weight of Antigen 37 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27163
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.