Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASAHL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116293
Description
ASAHL Polyclonal Antibody for Western Blot
N-acylsphingosine amidohydrolase-like 1 (ASAHL), commonly referred to as acid ceramidase-like protein, is a gene located on human chromosome 22q13.31. ASAHL encodes a protein that functions as a ceramidase, responsible for hydrolyzing ceramide into sphingosine and fatty acid, which are pivotal for cell signaling pathways that regulate cell growth, differentiation, and apoptosis. Though similar to acid ceramidase, ASAHL's specific biological role and tissue distribution remain active research areas, suggesting potential involvement in lipid metabolism and cellular homeostasis. Dysregulation of ceramidase activity, including ASAHL, is linked to various diseases, such as cancer, neurodegenerative disorders, and metabolic syndromes, due to the crucial role of sphingolipid signaling in these conditions. The gene's activity may influence ceramide levels within cells, impacting susceptibility to apoptosis and other cell fate decisions, highlighting its importance in pathophysiological contexts.
Specifications
| ASAHL | |
| Polyclonal | |
| Unconjugated | |
| Naaa | |
| 2210023K21Rik; 3830414F09Rik; acid ceramidase-like protein; Asahl; ASAH-like protein; LOW QUALITY PROTEIN: N-acylethanolamine-hydrolyzing acid amidase; Naaa; N-acylethanolamine acid amidase; N-acylethanolamine-hydrolyzing acid amidase; N-acylethanolamine-hydrolyzing acid amidase subunit alpha; N-acylethanolamine-hydrolyzing acid amidase subunit beta; N-acylsphingosine amidohydrolase-like; PLT | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 27163 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q02083 | |
| Naaa | |
| KLH conjugated synthetic peptide between 331-359 amino acids from the C-terminal region of human NAAA. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction