Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASAM Antibody (001), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290634AF350
Description
ASAM Monoclonal specifically detects ASAM in Mouse samples. It is validated for ELISA.Specifications
ASAM | |
Monoclonal | |
Alexa Fluor 350 | |
ACAMCoxsackie- and adenovirus receptor-like membrane protein, Adipocyte adhesion molecule, adipocyte-specific adhesion molecule, ASAMCAR-like membrane protein, CXADR-like membrane protein, FLJ22415 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ASAM (Q8R373-1; Met 1-Met 232). | |
0.1 mL | |
Signal Transduction | |
79827 | |
Mouse | |
Purified |
ELISA | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human ASAM | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction