Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASAM Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | ASAM |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5122225
|
Invitrogen™
PA5122225 |
20 μL |
Each for $237.00
|
|
|||||
PIPA5122224
![]() |
Invitrogen™
PA5122224 |
100 μL |
Each for $539.50
|
|
|||||
Description
May be involved in the cell-cell adhesion. May play a role in adipocyte differentiation and development of obesity. Is required for normal small intestine development.Specifications
ASAM | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9H6B4 | |
79827 | |
Thr19-Ser183. | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Clmp |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
9030425E11Rik; ACAM; Adipocyte adhesion molecule; adipocyte-specific adhesion molecule; adipocyte-specific protein 5; Asam; ASP5; AW557819; CAR-like membrane protein; Clmp; Coxsackie- and adenovirus receptor-like membrane protein; CSBM; CSBS; CXADR like membrane protein; CXADR-like membrane protein; FLJ22415; LOW QUALITY PROTEIN: CXADR-like membrane protein; Ol16; UNQ318/PRO363; visceral adipose tissue-specific transmembrane protein OL-16 | |
Clmp | |
IgG | |
Antigen affinity chromatography, Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title