Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119159
Description
ASAP1 Polyclonal specifically detects ASAP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ASAP1 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
130 kDa phosphatidylinositol 4,5-biphosphate-dependent ARF1 GTPase-activatingprotein, AMAP1, ARF GTPase-activating protein 1, arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 1, ArfGAP with SH3 domain, ankyrin repeat and PH domain 1, centaurin, beta 4, CENTB4, DDEF1ADP-ribosylation factor-directed GTPase-activating protein 1, development and differentiation enhancing factor 1, Development and differentiation-enhancing factor 1, Differentiation-enhancing factor 1, KIAA1249DEF-1, PAG2, PAP, PIP2-dependent ARF1 GAP, ZG14P | |
Rabbit | |
Affinity Purified | |
RUO | |
50807 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
NP_060952.2 | |
ASAP1 | |
The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human ArfGAP with SH3 domain, ankyrin repeat and PH domain 1 using the numbering given in entry NP_060952.2 (GeneID 50807). | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction