Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561770 Supplier Invitrogen™ Supplier No. PA561770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKPGELPPKP QLGDLPPKPQ LSDLPPKPQM KDLPPKPQLG DLLAKSQTGD VSPKAQQPSE VTLKSHPLDL SPNVQSRDAI QKQASEDSND LTPTLPETPV PLPRKINTGK NKVRRVKTIY DC Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

ASAP1, also known as Development and differentiation enhancing factor 1, is a protein with two ANK repeats, an Arf-GAP domain, a PH domain and a SH3 domain. ASAP1 is a homodimer known to interact with SRC and CRK and its activity is stimulated by phosphatidylinositol 4,5-biphosphate (PIP2). It possesses phosphatidylinositol 4,5-biphosphate-dependent GTPase-activating protein activity for ARF1 (ADP ribosylation factor 1) and ARF5 and a lesser activity towards ARF6. It may coordinate membrane trafficking with cell growth or actin cytoskeleton remodeling by binding to both SRC and PIP2 and may also function as a signal transduction protein involved in the differentiation of fibroblasts into adipocytes and possibly other cell types. It is widely expressed in most of the tissues. ASAP1 is a protooncogene involved in invasive breast cancer and in other types of invasive and malignant tumors, such as uveal melanomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASAP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASAP1
Gene Accession No. Q9ULH1
Gene Alias 130 kDa phosphatidylinositol 4,5-biphosphate-dependent ARF1 GTPase-activating protein; 130 kDa phosphatidylinositol 4,5-bisphosphate-dependent ARF1 GTPase-activating protein; ADP-ribosylation factor-directed GTPase-activating protein 1; AMAP1; ARF GTPase-activating protein 1; arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain1; Asap1; AV239055; centaurin, beta 4; CENTB4; Ddef1; DEF-1; development and differentiation enhancing factor 1; development and differentiation-enhancing factor 1; differentiation enhancing factor 1; differentiation-enhancing factor 1; Kiaa1249; mKIAA1249; PAG2; PAP; PIP2-dependent ARF1 GAP; s19; Shag1; src SH3 binding protein; ZG14P
Gene Symbols ASAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASAP1. Recombinant protein control fragment (Product #RP-104260).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50807
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.