Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASAP1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA125512
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA125512 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA125512 Supplier Thermo Scientific Supplier No. PA125512

Rabbit Polyclonal Antibody

Recommended positive controls: rat brain, mouse brain, NIH3T3. PA1-25512 is expected to cross react with bovine due to sequence homology. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ASAP1 Polyclonal antibody specifically detects ASAP1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASAP1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 11.6 mg/mL
Conjugate Unconjugated
Formulation 0.01M PBS with 15mM sodium azide; pH 7.4
Gene ASAP1
Gene Accession No. O97902, Q9ULH1, Q9QWY8, Q1AAU6
Gene Alias 130 kDa phosphatidylinositol 4,5-biphosphate-dependent ARF1 GTPase-activating protein; 130 kDa phosphatidylinositol 4,5-bisphosphate-dependent ARF1 GTPase-activating protein; ADP-ribosylation factor-directed GTPase-activating protein 1; AMAP1; ARF GTPase-activating protein 1; arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain1; Asap1; AV239055; centaurin, beta 4; CENTB4; Ddef1; DEF-1; development and differentiation enhancing factor 1; development and differentiation-enhancing factor 1; differentiation enhancing factor 1; differentiation-enhancing factor 1; Kiaa1249; mKIAA1249; PAG2; PAP; PIP2-dependent ARF1 GAP; s19; Shag1; src SH3 binding protein; ZG14P
Gene Symbols ASAP1
Host Species Rabbit
Immunogen Synthetic peptide, conjugated to KLH, corresponding to residues 1128-1147 of mouse Centaurin beta 4. The peptide sequence is identical in bovine ASAP1
Purification Method IgG fraction
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13196, 314961, 327705, 50807
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.