Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144952 Supplier Invitrogen™ Supplier No. PA5144952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WRLLHEDLDE SDDDMDEKLQ PSPNRREDRP ISFYQLGSNQ LQSNAVSLAR DAANLAKDKQ RAFMPSILQN ETY Antigen sequence identity: Mouse 89%; Rat 88%.

ASAP2 is a multidomain protein belonging to the beta family with two ANK repeats, an Arf-GAP domain, a PH domain and a SH3 domain. This protein localizes in the Golgi apparatus and also at the plasma membrane where it colocalizes with protein tyrosine kinase 2-beta (PYK2) and forms a stable complex with PYK2 in vivo. ASAP2 is tyrosine phosphorylated by activated PYK2. It functions as a substrate and downstream target for PYK2 and SRC, a pathway that may be involved in the regulation of vesicular transport. ASAP2 activates the small GTPases ARF1, ARF5 and ARF6 and regulates the formation of post-Golgi vesicles and modulates constitutive secretion. It modulates phagocytosis mediated by Fc gamma receptor and ARF6. Usually detected in heart, brain, placenta, kidney, monocytes and pancreas, it is up-regulated during monocyte maturation. Multiple transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASAP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASAP2
Gene Accession No. O43150
Gene Alias 6530401G17Rik; AMAP2; Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2; ArfGAP with SH3 domain, ankyrin repeat and PH domain 2; Asap2; centaurin, beta 3; CENTB3; DDEF2; development and differentiation enhancing factor 2; development and differentiation-enhancing factor 2; Gm1523; Gm592; KIAA0400; PAG3; PAP; Pap-alpha; PAPbeta; Paxillin-associated protein with ARF GAP activity 3; PYK2 C terminus-associated protein; pyk2 C-terminus-associated protein; SHAG1
Gene Symbols ASAP2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 691-763. Recombinant protein control fragment (Product #RP-109959).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.