Learn More
ASAP2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303311A
Description
The recommended shelf life for this product is 1 year from date of receipt.
ASAP2 is a multidomain protein belonging to the beta family with two ANK repeats, an Arf-GAP domain, a PH domain and a SH3 domain. This protein localizes in the Golgi apparatus and also at the plasma membrane where it colocalizes with protein tyrosine kinase 2-beta (PYK2) and forms a stable complex with PYK2 in vivo. ASAP2 is tyrosine phosphorylated by activated PYK2. It functions as a substrate and downstream target for PYK2 and SRC, a pathway that may be involved in the regulation of vesicular transport. ASAP2 activates the small GTPases ARF1, ARF5 and ARF6 and regulates the formation of post-Golgi vesicles and modulates constitutive secretion. It modulates phagocytosis mediated by Fc gamma receptor and ARF6. Usually detected in heart, brain, placenta, kidney, monocytes and pancreas, it is up-regulated during monocyte maturation. Multiple transcript variants encoding different isoforms have been found.Specifications
| ASAP2 | |
| Polyclonal | |
| Unconjugated | |
| ASAP2 | |
| AMAP2, centaurin, beta 3, CENTB3, DDEF2, development and differentiation enhancing factor 2, development and differentiation-enhancing factor 2, KIAA0400, PAG3, PAP, Pap-alpha, paxillin-associated protein with ARF GAP activity 3, PYK2 C terminus-associated protein, pyk2 C-terminus-associated protein, SHAG1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8853 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O43150 | |
| ASAP2 | |
| Between 900 and 950 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.