Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582729 Supplier Invitrogen™ Supplier No. PA582729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKHHKECEEL LEQAQAGTFA FPLHVDYSWV ISTEPGSDSE EDEEEKRCLL KLPAQAHWAS GRLDISNKTY ETVASLGAAT PQGESEDCPP PLPVKNSSRT LVQGCAR Highest antigen sequence indentity to the following orthologs: Rat - 66%, Mouse - 70%.

This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASAP3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Asap3
Gene Accession No. Q8TDY4
Gene Alias 9430088F20Rik; ACAP4; ARF6 GTPase-activating protein; Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 3; ArfGAP with SH3 domain, ankyrin repeat and PH domain 3; ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 1; Asap3; centaurin, beta 6; CENTB6; DDEFL1; development and differentiation enhancing factor-like 1; development and differentiation-enhancing factor-like 1; Gm140; Protein up-regulated in liver cancer 1; UPLC1; up-regulated in liver cancer 1 (UPLC1)
Gene Symbols Asap3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASAP3. Recombinant protein control fragment (Product #RP-91723).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55616
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.