Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570658
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 93%.
The protein encoded by this gene contains an ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signaling (SOCS) proteins and their binding partners with the elongin B and C complex, targeting them for ubiquitination and degradation.
Specifications
ASB1 | |
Polyclonal | |
Unconjugated | |
ASB1 | |
1700029O08Rik; 1700054C17Rik; AI426616; AI851378; ankyrin repeat and SOCS box containing 1; ankyrin repeat and SOCS box protein 1; ankyrin repeat and SOCS box-containing 1; ankyrin repeat and SOCS box-containing protein 1; ankyrin repeat-containing SOCS box protein 1; ASB1; ASB-1; KIAA1146; LOW QUALITY PROTEIN: ankyrin repeat and SOCS box protein 1; mKIAA1146 | |
Rabbit | |
Affinity chromatography | |
RUO | |
51665 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y576 | |
ASB1 | |
synthetic peptide directed towards the middle region of human ASB1 (aa 236-285). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction