Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551946 Supplier Invitrogen™ Supplier No. PA551946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVDVKGQTAL YVAVVNGHLE STQILLEAGA DPNGSRHHRS TPVYHASRVG RADILKALIR YGADVDVNHH LTPDVQPRFS RRLTSLVVCP LYISAAYHNL QCFRLLLLAG ANPDFNCNGP VNTQGFYRGS PGCVMDAVL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

The protein encoded by this gene contains an ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signaling (SOCS) proteins and their binding partners with the elongin B and C complex, targeting them for ubiquitination and degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASB1
Gene Accession No. Q9Y576
Gene Alias 1700029O08Rik; 1700054C17Rik; AI426616; AI851378; ankyrin repeat and SOCS box containing 1; ankyrin repeat and SOCS box protein 1; ankyrin repeat and SOCS box-containing 1; ankyrin repeat and SOCS box-containing protein 1; ankyrin repeat-containing SOCS box protein 1; ASB1; ASB-1; KIAA1146; LOW QUALITY PROTEIN: ankyrin repeat and SOCS box protein 1; mKIAA1146
Gene Symbols ASB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASB1. Recombinant protein control fragment (Product #RP-102064).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51665
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.