Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA129820
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA129820 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA129820 Supplier Invitrogen™ Supplier No. PA129820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-29820 detects ASB11 from human samples.

The ankyrin repeat and SOCS box-containing (Asb) protein family is comprised of members which each contain 'SOCS boxes' but differ from other families of protein in the motifs they contain upstream of the SOCS box (1). SOCS boxes are carboxy-terminal regions of homology found in the suppressor of cytokine signaling family of proteins. The box region is thought to be the point of interaction between SOCS proteins and E3 ubiquitin ligases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% BSA and 0.02% sodium azide
Gene ASB11
Gene Accession No. Q8WXH4
Gene Alias 1110067L12Rik; 1600009D24Rik; ankyrin repeat and SOCS box containing 11; ankyrin repeat and SOCS box containing 11, E3 ubiquitin protein ligase; ankyrin repeat and SOCS box containing protein a; ankyrin repeat and SOCS box protein 11; ankyrin repeat and SOCS box-containing 11; ankyrin repeat and SOCS box-containing protein 11; ankyrin repeat domain-containing SOCS box protein ASB11; Asb11; ASB-11; asb-a; asb-a protein; d-asb11; DKFZp779E2460; MGC119168; MGC119169; RGD1566298; zgc:136370; zgc:158532
Gene Symbols ASB11
Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids 40-51 and 250-265 of human Asb-11.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140456
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.