Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562002 Supplier Invitrogen™ Supplier No. PA562002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLMDITKIFS LLQPDKEEED TDTEEKQALN QAVYDNDSYT LDQLLRQERY KRFINSRSGW GVPGTPLRLA ASYGHLSCLQ VLL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. The SOCS box domain has not yet been annotated for this encoded protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASB12
Gene Accession No. Q8WXK4
Gene Alias ankyrin repeat and SOCS box containing 12; ankyrin repeat and SOCS box protein 12; ankyrin repeat and SOCS box-containing 12; ankyrin repeat domain-containing SOCS box protein Asb-12; ASB12; ASB-12
Gene Symbols ASB12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASB12. Recombinant protein control fragment (Product #RP-104453).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142689
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.