Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASB12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107068
Description
Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. The SOCS box domain has not yet been annotated for this encoded protein.
Specifications
| ASB12 | |
| Polyclonal | |
| Unconjugated | |
| ASB12 | |
| ankyrin repeat and SOCS box containing 12; ankyrin repeat and SOCS box protein 12; ankyrin repeat and SOCS box-containing 12; ankyrin repeat domain-containing SOCS box protein Asb-12; ASB12; ASB-12 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 142689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q8WXK4 | |
| ASB12 | |
| Synthetic peptide directed towards the C-terminal region of human ASB12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction