Learn More
ASB13 Mouse anti-Human, Clone: OTI3B2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809440

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing family of proteins. They contain ankyrin repeat sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length sequences are not known.Specifications
ASB13 | |
Monoclonal | |
Unconjugated | |
ASB13 | |
ankyrin repeat and SOCS box containing 13 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79754 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3B2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WXK3 | |
ASB13 | |
Human recombit protein fragment corresponding to amino acids 76-278 of human ASB13 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.