Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590802 Supplier Invitrogen™ Supplier No. PA590802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEPRAADGCF LGDVGFWVER TPVHEAAQRG ESLQLQQLIE SGACVNQVTV DSITPLHAAS LQGQARCVQL LLAAGAQVDA RNIDGSTPLC DACASGSIEC VKLLLSYGAK VNPPLYTASP LHEACMSGSS ECVRLLIDVG ANLEAHDCHF GTPLHVACAR EHLDCVKVLL NAGANVNAAK LHETALHHAA KVKNVDLIEM LIEFGGNIYA RDNRGKKPSD YTWSSSAPAK CFEYYEKTPL TLSQLCRVNL RKATGVRGLE KIAKLNIPPR LIDYLSYN Positive Samples: Mouse kidney.

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing family of proteins. They contain ankyrin repeat sequence and a SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length sequences are not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB13
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ASB13
Gene Accession No. Q8VBX0, Q8WXK3
Gene Alias 2210015B19Rik; 6430573K02Rik; ankyrin repeat and SOCS box containing 13; ankyrin repeat and SOCS box protein 13; ankyrin repeat and SOCS box-containing 13; ankyrin repeat and SOCS box-containing protein 13; ankyrin repeat domain-containing SOCS box protein 13; ankyrin repeat domain-containing SOCS box protein Asb-13; ASB13; Asb-13; C85285
Gene Symbols ASB13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-278 of human ASB13 (NP_0789772).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142688, 361268, 79754
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.