Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASB14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570812
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 86%.
ASB14 gene ontology annotations related to this gene include intracellular signal transduction; protein ubiquitination.
Specifications
ASB14 | |
Polyclonal | |
Unconjugated | |
ASB14 | |
ankyrin repeat and SOCS box containing 14; ankyrin repeat and SOCS box protein 14; ankyrin repeat and SOCS box-containing 14; Ankyrin repeat and SOCS box-containing protein 14; ankyrin repeat domain-containing SOCS box protein 14; ankyrin repeat domain-containing SOCS box protein Asb-14; ASB14; ASB-14; Axonemal beta dynein heavy chain 12; ciliary dynein heavy chain 12; DNAH12; Dynein heavy chain 12, axonemal | |
Rabbit | |
Affinity chromatography | |
RUO | |
142686 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A6NK59 | |
ASB14 | |
synthetic peptide directed towards the N-terminal region of human ASB14. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction