Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116948
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116948 Supplier Invitrogen™ Supplier No. PA5116948
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: LNCaP, Rat testis Immunogen sequence: GHRFELNFNL EFTEICVNTI LYWVFARKGN PDFVELLLKK TKDYVQDRSC SLALIWRTFT PVYCPSPLSG ITPLLYVAQT RQSNILKILL QYGILEREKN.

May be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB17
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.656 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ASB17
Gene Accession No. Q8WXJ9
Gene Alias 4933436O18Rik; ankyrin repeat and SOCS box containing 17; ankyrin repeat and SOCS box containing protein 17; ankyrin repeat and SOCS box protein 17; ankyrin repeat and SOCS box-containing 17; ankyrin repeat and SOCS box-containing protein 17; Asb17; ASB-17
Gene Symbols ASB17
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 78-177 of mouse ASB17 (NP_0800342).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127247, 687364
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.