Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564370 Supplier Invitrogen™ Supplier No. PA564370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DYLHDYPLNS DLVKRLKSAL DAKDEERVRD LICTEITPVD AVIELANDDW MKDPSAQLPT GMLLGDLD Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 71%.

ASB18 may be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASB18
Gene Accession No. Q6ZVZ8
Gene Alias ankyrin repeat and SOCS box containing 18; ankyrin repeat and SOCS box protein 18; ankyrin repeat and SOCS box-containing 18; ASB18; ASB-18
Gene Symbols ASB18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASB18. Recombinant protein control fragment (Product #RP-103942).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401036
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.