Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB2 Monoclonal Antibody (OTI2A2)
GREENER_CHOICE

Catalog No. PIMA526939
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526939 Supplier Invitrogen™ Supplier No. MA526939
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ASB2 Monoclonal Antibody for IHC (P)

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. This gene is induced by all-trans retinoic acid. In myeloid leukemia cells, the expression of this encoded protein has been shown to induce growth inhibition and chromatin condensation. Multiple alternatively spliced transcript variants have been described for this gene but their full length sequences are not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ASB2
Gene Accession No. Q96Q27
Gene Alias 1110008E15Rik; ankyrin repeat and SOCS box containing 2; ankyrin repeat and SOCS box protein 2; ankyrin repeat and SOCS box-containing 2; ankyrin repeat and SOCS box-containing protein 2; ankyrin repeat and SOCS box-containing protein 2a; ankyrin repeat-containing SOCS box protein 2; Asb2; ASB-2; asb2 {ECO:0000312; EMBL:AAH85882.1}; MGC40044
Gene Symbols ASB2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-300 of ASB2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51676
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.