Learn More
Invitrogen™ ASB2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ASB2 protein.
Specifications
Specifications
| Antigen | ASB2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ASB2 |
| Gene Accession No. | Q96Q27 |
| Gene Alias | 1110008E15Rik; ankyrin repeat and SOCS box containing 2; ankyrin repeat and SOCS box protein 2; ankyrin repeat and SOCS box-containing 2; ankyrin repeat and SOCS box-containing protein 2; ankyrin repeat and SOCS box-containing protein 2a; ankyrin repeat-containing SOCS box protein 2; Asb2; ASB-2; asb2 {ECO:0000312; EMBL:AAH85882.1}; MGC40044 |
| Gene Symbols | ASB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.