Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASB3 Monoclonal Antibody (3E5)
GREENER_CHOICE

Catalog No. PIMA521864
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521864 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521864 Supplier Invitrogen™ Supplier No. MA521864
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MDFTEAYADT CSTVGLAARE GNVKVLRKLL KKGRSVDVAD NRGWMPIHEA AYHNSVECLQ MLINADSSEN YIKMKTFEGF CALHLAASQG HWKIVQILLE AGADPNATTL EETTPLFLAV ENGQIDVLRL LLQHGANVNG SHSMCGWNSL HQASFQENAE IIKLLLRKGA NKECQDDFGI TPLFVAAQYG KLESLSILIS SGANVNCQAL DKATPLFIAA QEGHTKCVEL LLSSGADPDL YCNEDSWQLP IHAAAQMGHT KILDLLIPLT NRACDTGLNK VSPVYSAVFG GHEDCLEILL RNGYSPDAQA CLVFGFSSPV CMAFQKDCEF FGIVNILLKY GAQINELHLA YCLKYEKFSI FRYFLRKGCS LGPWNHIYEF VNHAIKAQAK YKEWLPHLLV AGFDPLILLC NSWIDSVSID TPIFTLEFTN WKTLAPAVER MLSARASNAW ILQQHIATVP SLTHLCRLEI RSSLKSERLR SDSYISQLPL PRSLHNYLLY EDVLRMYEVP ELAAIQDG.

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. Multiple alternatively spliced transcript variants have been described for this gene but some of the full length sequences are not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB3
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3E5
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ASB3
Gene Accession No. Q9Y575
Gene Alias 2400011J03Rik; ankyrin repeat and SOCS box containing 3; ankyrin repeat and SOCS box protein 3; ankyrin repeat and SOCS box-containing 3; ankyrin repeat and SOCS box-containing 3-like; ankyrin repeat and SOCS box-containing protein 3; ankyrin repeat-containing SOCS box protein 3; ASB3; Asb-3; EAR1; FLJ10123; FLJ10421; HREV; MGC12531; MGC132002; MGC996; THRAL
Gene Symbols ASB3
Host Species Mouse
Immunogen ASB3 (AAH06488.1, 1 a.a. ~ 518 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51130
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.